Assay Details
Target Gene Details
Entrez Gene ID: | 11143 |
Gene Name: | lysine acetyltransferase 7 |
Gene Aliases: |
HBO1, HBOA, MYST2, ZC2HC7 |
Location: |
Chr.17:49788619-49829096 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KAT7 | NM_001199155.1 | NP_001186084.1 | ||
| NM_001199156.1 | NP_001186085.1 | |||
| NM_001199157.1 | NP_001186086.1 | |||
| NM_001199158.1 | NP_001186087.1 | |||
| NM_007067.4 | NP_008998.1 | |||
| AF074606.1 | AAC99368.1 | |||
| AF140360.1 | AAD42348.1 | |||
| AF217502.1 | AAL56649.1 | |||
| AK023890.1 | ||||
| AK122932.1 | ||||
| AK293976.1 | ||||
| AK294014.1 | ||||
| AK294052.1 | ||||
| AK294836.1 | ||||
| AK297212.1 | ||||
| BC032640.2 | AAH32640.1 |
More Information
Set Membership: |
|

Genomic Map