Assay Details
Target Gene Details
Entrez Gene ID: | 1353 |
Gene Name: | COX11, cytochrome c oxidase copper chaperone |
Gene Aliases: |
COX11P |
Location: |
Chr.17:54951898-54971024 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COX11 | NM_001162861.2 | NP_001156333.1 | ||
| NM_001162862.2 | NP_001156334.1 | |||
| NM_001321518.1 | NP_001308447.1 | |||
| NM_004375.4 | NP_004366.1 | |||
| NR_027941.2 | ||||
| NR_027942.2 | ||||
| NR_135677.1 | ||||
| XM_011524342.2 | XP_011522644.1 | |||
| XM_017024192.1 | XP_016879681.1 | |||
| XM_017024193.1 | XP_016879682.1 | |||
| XM_017024194.1 | XP_016879683.1 | |||
| XM_017024195.1 | XP_016879684.1 | |||
| XM_017024196.1 | XP_016879685.1 | |||
| AF044321.1 | AAD08645.1 | |||
| AK293851.1 | ||||
| AK295381.1 | ||||
| BC005895.1 | AAH05895.1 | |||
| BF725369.1 | ||||
| BI598312.1 | ||||
| BM975167.1 | ||||
| CR541837.1 | CAG46636.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv3266n100 | Chr.17:54817463 - 55002289 on Build GRCh38 | Loss |
|
| nsv457853 | Chr.17:54910871 - 54982748 on Build GRCh38 | Loss |
|
| dgv5708n54 | Chr.17:54962122 - 55083585 on Build GRCh38 | Loss |
|
| esv3640830 | Chr.17:54919823 - 54998005 on Build GRCh38 | Gain |
|
| nsv522299 | Chr.17:54966408 - 55196549 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map