Assay Details
Target Gene Details
Entrez Gene ID: | 54828 |
Gene Name: | BCAS3, microtubule associated cell migration factor |
Gene Aliases: |
GAOB1, MAAB |
Location: |
Chr.17:60677811-61392838 on Build GRCh38 |
Assay Gene Location: | Within Intron 34 |
Target Gene Details
Entrez Gene ID: | 101927855 |
Gene Name: | uncharacterized LOC101927855 |
Gene Aliases: |
- |
Location: |
Chr.17:61361668-61367704 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101927855 | NR_136397.1 | |||
| BX115170.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv43n68 | Chr.17:61246105 - 61485623 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map