Assay Details
Target Gene Details
Entrez Gene ID: | 3588 |
Gene Name: | interleukin 10 receptor subunit beta |
Gene Aliases: |
CDW210B, CRF2-4, CRFB4, D21S58, D21S66, IL-10R2 |
Location: |
Chr.21:33266360-33297234 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IL10RB | NM_000628.4 | NP_000619.3 | ||
| AK124057.1 | BAC85761.1 | |||
| AK299961.1 | ||||
| AK313348.1 | ||||
| AU133530.2 | ||||
| BC001903.1 | AAH01903.1 | |||
| BT009777.1 | AAP88779.1 | |||
| JN591378.1 | ||||
| Z17227.1 | CAA78933.1 |
More Information
Set Membership: |
|

Genomic Map