Assay Details
Target Gene Details
Entrez Gene ID: | 2618 |
Gene Name: | phosphoribosylglycinamide formyltransferase, phosphoribosylglycinamide synthetase, phosphoribosylaminoimidazole synthetase |
Gene Aliases: |
AIRS, GARS, GARTF, PAIS, PGFT, PRGS |
Location: |
Chr.21:33503931-33542917 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GART | NM_000819.4 | NP_000810.1 | ||
| NM_001136005.1 | NP_001129477.1 | |||
| NM_001136006.1 | NP_001129478.1 | |||
| NM_175085.2 | NP_780294.1 | |||
| XM_005260941.1 | XP_005260998.1 | |||
| XM_006723989.1 | XP_006724052.1 | |||
| XM_006723990.1 | XP_006724053.1 | |||
| XM_011529526.1 | XP_011527828.1 | |||
| AB208785.1 | BAD92022.1 | |||
| AK292560.1 | ||||
| AK292897.1 | ||||
| BC038958.1 | AAH38958.1 | |||
| BC093641.1 | AAH93641.1 | |||
| BC101565.1 | AAI01566.1 | |||
| BC107712.1 | AAI07713.1 | |||
| BX648124.1 | ||||
| D32051.1 | BAA06809.1 | |||
| X54199.1 | CAA38119.1 |
More Information
Set Membership: |
|

Genomic Map