Assay Details
Target Gene Details
Entrez Gene ID: | 522 |
Gene Name: | ATP synthase, H+ transporting, mitochondrial Fo complex subunit F6 |
Gene Aliases: |
ATP5, ATP5A, ATPM, CF6, F6 |
Location: |
Chr.21:25724480-25735654 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATP5J | NM_001003696.1 | NP_001003696.1 | ||
| NM_001003697.1 | NP_001003697.1 | |||
| NM_001003701.1 | NP_001003701.1 | |||
| NM_001003703.1 | NP_001003703.1 | |||
| NM_001320266.1 | NP_001307195.1 | |||
| NM_001320267.1 | NP_001307196.1 | |||
| NM_001685.4 | NP_001676.2 | |||
| AA364611.1 | ||||
| AK311899.1 | ||||
| AL110183.1 | CAB53667.1 | |||
| AY544129.1 | AAT11160.1 | |||
| BC001178.1 | AAH01178.1 | |||
| BC066310.1 | AAH66310.1 | |||
| BG938438.1 | ||||
| BQ268216.1 | ||||
| BU174560.1 | ||||
| CN260182.1 | ||||
| HY064673.1 | ||||
| HY146471.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1064561 | Chr.21:25714853 - 25750046 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map