Assay Details
Target Gene Details
Entrez Gene ID: | 100874093 |
Gene Name: | BRWD1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.21:39315707-39323218 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BRWD1-AS1 | NR_046655.1 | |||
| DB057251.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv587480 | Chr.21:39199320 - 39329243 on Build GRCh38 | Loss |
|
| nsv587482 | Chr.21:39312501 - 39334668 on Build GRCh38 | Loss |
|
| nsv587483 | Chr.21:39314401 - 39346235 on Build GRCh38 | Loss |
|

Genomic Map