Assay Details
Target Gene Details
Entrez Gene ID: | 116448 |
Gene Name: | oligodendrocyte transcription factor 1 |
Gene Aliases: |
BHLHB6, BHLHE21 |
Location: |
Chr.21:33070144-33072422 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OLIG1 | NM_138983.2 | 1 | 111 | NP_620450.2 |
| AB209791.1 | 1 | 102 | BAD93028.1 | |
| AL834450.1 | 1 | 104 | CAD39110.1 | |
| BC026989.1 | AAH26989.1 | |||
| BC033290.1 | 1 | 118 | AAH33290.1 |
More Information
Set Membership: |
|

Genomic Map