Assay Details
Target Gene Details
Entrez Gene ID: | 29104 |
Gene Name: | N-6 adenine-specific DNA methyltransferase 1 (putative) |
Gene Aliases: |
C21orf127, HEMK2, MTQ2, N6AMT, PRED28, m.HsaHemK2P |
Location: |
Chr.21:28571095-28885373 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| N6AMT1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv275545 | Chr.21:28601975 - 28605933 on Build GRCh38 | Loss |
|

Genomic Map