Assay Details
Target Gene Details
Entrez Gene ID: | 100506215 |
Gene Name: | PAXBP1 antisense RNA 1 |
Gene Aliases: |
GCFC1-AS1 |
Location: |
Chr.21:32728114-32743122 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PAXBP1-AS1 | NR_038879.1 | 1 | 620 | |
| NR_038880.1 | 1 | 620 | ||
| AK056384.1 | 1 | 619 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3646891 | Chr.21:32675352 - 32751209 on Build GRCh38 | Gain |
|
| nsv834086 | Chr.21:32710449 - 32883526 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs139940454] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map