Assay Details
Target Gene Details
Entrez Gene ID: | 2551 |
Gene Name: | GA binding protein transcription factor alpha subunit |
Gene Aliases: |
E4TF1-60, E4TF1A, NFT2, NRF2, NRF2A, RCH04A07 |
Location: |
Chr.21:25734947-25772460 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GABPA | NM_001197297.1 | NP_001184226.1 | ||
| NM_002040.3 | NP_002031.2 | |||
| XM_005260938.4 | XP_005260995.1 | |||
| XM_011529520.2 | XP_011527822.1 | |||
| XM_011529521.2 | XP_011527823.1 | |||
| XM_017028313.1 | XP_016883802.1 | |||
| AK223164.1 | BAD96884.1 | |||
| AK313545.1 | ||||
| BC035031.2 | AAH35031.1 | |||
| BX647755.1 | ||||
| D13318.1 | BAA02575.1 | |||
| U13044.1 | AAA65706.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1064561 | Chr.21:25714853 - 25750046 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs71649667] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map