Assay Details
Target Gene Details
Entrez Gene ID: | 25776 |
Gene Name: | chibby family member 1, beta catenin antagonist |
Gene Aliases: |
C22orf2, CBY, HS508I15A, PGEA1, PIGEA-14, PIGEA14, arb1 |
Location: |
Chr.22:38656653-38673850 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CBY1 | NM_001002880.1 | NP_001002880.2 | ||
| NM_015373.3 | NP_056188.1 | |||
| AB015347.1 | BAA88119.1 | |||
| AB111855.1 | BAC78839.1 | |||
| AF331041.1 | AAL56062.1 | |||
| AK307265.1 | ||||
| AK311928.1 | ||||
| AL050345.1 | CAB43547.1 | |||
| AL136686.1 | CAB66621.1 | |||
| BC016139.1 | AAH16139.1 | |||
| CD702742.1 | ||||
| CR456410.1 | CAG30296.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv508733 | Chr.22:38627252 - 38684460 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map