Assay Details
Target Gene Details
Entrez Gene ID: | 402055 |
Gene Name: | SRR1 domain containing |
Gene Aliases: |
HC/HCC, SRR1L |
Location: |
Chr.22:26483867-26494620 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SRRD | NM_001013694.2 | NP_001013716.2 | ||
| XM_011530178.2 | XP_011528480.1 | |||
| XM_017028799.1 | XP_016884288.1 | |||
| AK293916.1 | ||||
| BC066962.1 | AAH66962.1 | |||
| DA325985.1 |
More Information
Set Membership: |
|

Genomic Map