Assay Details
Target Gene Details
Entrez Gene ID: | 5880 |
Gene Name: | ras-related C3 botulinum toxin substrate 2 (rho family, small GTP binding protein Rac2) |
Gene Aliases: |
EN-7, Gx, HSPC022, p21-Rac2 |
Location: |
Chr.22:37225261-37244299 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAC2 | NM_002872.4 | NP_002863.1 | ||
| XM_006724286.3 | XP_006724349.1 | |||
| AF498965.1 | AAM21112.1 | |||
| BC001485.2 | AAH01485.1 | |||
| BI908751.1 | ||||
| BQ934070.1 | ||||
| BT006919.1 | AAP35565.1 | |||
| CR456555.1 | CAG30441.1 | |||
| M29871.1 | AAA36538.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829203 | Chr.22:36998126 - 37773322 on Build GRCh38 | Loss |
|
| nsv834188 | Chr.22:37210658 - 37396179 on Build GRCh38 | Loss |
|
| nsv588987 | Chr.22:37215767 - 37242920 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map