Assay Details
Target Gene Details
Entrez Gene ID: | 9701 |
Gene Name: | protein phosphatase 6 regulatory subunit 2 |
Gene Aliases: |
KIAA0685, PP6R2, SAP190, SAPS2 |
Location: |
Chr.22:50343317-50445090 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv26897 | Chr.22:50343296 - 50348608 on Build GRCh38 | Loss |
|
| nsv589217 | Chr.22:50343738 - 50353791 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map