Assay Details
Target Gene Details
Entrez Gene ID: | 9402 |
Gene Name: | GRB2-related adaptor protein 2 |
Gene Aliases: |
GADS, GRAP-2, GRB2L, GRBLG, GRID, GRPL, GrbX, Grf40, Mona, P38 |
Location: |
Chr.22:39901073-39973343 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GRAP2 | NM_004810.3 | NP_004801.1 | ||
| XM_006724376.3 | XP_006724439.1 | |||
| AF042380.1 | AAC69273.1 | |||
| AF090456.1 | AAD13027.1 | |||
| AF102694.1 | AAD04926.1 | |||
| AF129476.1 | AAD41782.1 | |||
| AF236119.1 | AAF60319.1 | |||
| AJ011736.1 | CAA09757.1 | |||
| AK303332.1 | ||||
| AK303512.1 | ||||
| AY069959.1 | AAL58573.1 | |||
| BC025692.1 | AAH25692.1 | |||
| CR456498.1 | CAG30384.1 | |||
| Y18051.1 | CAA77021.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv510808 | Chr.22:39873227 - 39957952 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs139552253] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map