Assay Details
Target Gene Details
Entrez Gene ID: | 150379 |
Gene Name: | patatin like phospholipase domain containing 5 |
Gene Aliases: |
GS2L, dJ388M5, dJ388M5.4 |
Location: |
Chr.22:43879678-43892013 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PNPLA5 | NM_001177675.1 | NP_001171146.1 | ||
| NM_138814.3 | NP_620169.1 | |||
| AK056648.1 | ||||
| AK127172.1 | BAC86866.1 | |||
| BC031820.2 | ||||
| BC104839.1 | AAI04840.1 | |||
| BC104843.1 | AAI04844.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834219 | Chr.22:43853996 - 44012877 on Build GRCh38 | Gain |
|
| nsv1055831 | Chr.22:43878234 - 43932651 on Build GRCh38 | Gain |
|
| nsv829265 | Chr.22:43828073 - 43928999 on Build GRCh38 | Loss |
|
| nsv834218 | Chr.22:43800912 - 43982015 on Build GRCh38 | Loss |
|
| nsv834217 | Chr.22:43745292 - 43920182 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs116065637] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map