Assay Details
Target Gene Details
Entrez Gene ID: | 3560 |
Gene Name: | interleukin 2 receptor subunit beta |
Gene Aliases: |
CD122, IL15RB, P70-75 |
Location: |
Chr.22:37125838-37149922 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IL2RB | NM_000878.3 | NP_000869.1 | ||
| AK312860.1 | ||||
| BC025691.1 | AAH25691.1 | |||
| CR456506.1 | CAG30392.1 | |||
| M26062.1 | AAA59143.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834187 | Chr.22:36965603 - 37163260 on Build GRCh38 | Loss |
|
| nsv829203 | Chr.22:36998126 - 37773322 on Build GRCh38 | Loss |
|
| esv34015 | Chr.22:36966049 - 37189804 on Build GRCh38 | Loss |
|
| nsv588984 | Chr.22:36967923 - 37161398 on Build GRCh38 | Gain |
|
| nsv588986 | Chr.22:37133758 - 37145791 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map