Assay Details
Target Gene Details
Entrez Gene ID: | 23786 |
Gene Name: | BCL2 like 13 |
Gene Aliases: |
BCL-RAMBO, Bcl2-L-13, MIL1 |
Location: |
Chr.22:17628855-17730855 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BCL2L13 | XM_011546119.1 | XP_011544421.1 | ||
| XM_011546120.1 | XP_011544422.1 | |||
| XM_011546121.2 | XP_011544423.1 | |||
| XM_017028725.1 | XP_016884214.1 | |||
| XM_017028726.1 | XP_016884215.1 | |||
| XM_017028728.1 | XP_016884217.1 | |||
| XM_017028731.1 | XP_016884220.1 | |||
| XM_017028734.1 | XP_016884223.1 | |||
| AF246665.1 | AAK15150.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv9871 | Chr.22:17545048 - 17683541 on Build GRCh38 | Loss |
|
| nsv526826 | Chr.22:17411511 - 17836413 on Build GRCh38 | Gain |
|
| esv3893428 | Chr.22:17578563 - 18040448 on Build GRCh38 | Loss |
|
| esv3893429 | Chr.22:17600825 - 17678777 on Build GRCh38 | Gain |
|
| dgv4465n100 | Chr.22:17577691 - 17706530 on Build GRCh38 | Gain |
|
| nsv428385 | Chr.22:17491941 - 18886703 on Build GRCh38 | Gain+Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs139504598] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map