Assay Details
Target Gene Details
Entrez Gene ID: | 27124 |
Gene Name: | inositol polyphosphate-5-phosphatase J |
Gene Aliases: |
INPP5, PIB5PA, PIPP |
Location: |
Chr.22:31107736-31134697 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| INPP5J | XM_011530143.1 | XP_011528445.1 | ||
| XM_017028771.1 | XP_016884260.1 | |||
| XM_017028773.1 | XP_016884262.1 | |||
| AK308921.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv964621 | Chr.22:31107865 - 31113534 on Build GRCh38 | Duplication |
|
| nsv829173 | Chr.22:31054057 - 31147970 on Build GRCh38 | Loss |
|
| nsv9896 | Chr.22:30973218 - 31117947 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map