Assay Details
Target Gene Details
Entrez Gene ID: | 5988 |
Gene Name: | ret finger protein like 1 |
Gene Aliases: |
RNF78 |
Location: |
Chr.22:29403031-29442491 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RFPL1 | XM_011530298.2 | XP_011528600.1 | ||
| XM_017028900.1 | XP_016884389.1 | |||
| XM_017028901.1 | XP_016884390.1 | |||
| XM_017028902.1 | XP_016884391.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1059994 | Chr.22:29401384 - 29438777 on Build GRCh38 | Gain |
|
| nsv834171 | Chr.22:29230413 - 29433855 on Build GRCh38 | Loss |
|
| nsv1060197 | Chr.22:29421518 - 29446522 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map