Assay Details
Target Gene Details
Entrez Gene ID: | 27254 |
Gene Name: | cold shock domain containing C2 |
Gene Aliases: |
PIPPIN, dJ347H13.2 |
Location: |
Chr.22:41561010-41576666 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CSDC2 | NM_014460.3 | NP_055275.1 | ||
| AB027011.1 | BAA84704.1 | |||
| AK313407.1 | ||||
| AL834417.1 | CAD39079.2 | |||
| CR456457.1 | CAG30343.1 | |||
| DB168570.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv516925 | Chr.22:41252498 - 41585953 on Build GRCh38 | Loss |
|
| nsv1055166 | Chr.22:41536399 - 41622240 on Build GRCh38 | Gain |
|
| nsv834209 | Chr.22:41563516 - 41746056 on Build GRCh38 | Loss |
|
| nsv524585 | Chr.22:40993505 - 41794305 on Build GRCh38 | Loss |
|
| nsv3639 | Chr.22:41571809 - 41602871 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map