Assay Details
Target Gene Details
Entrez Gene ID: | 25817 |
Gene Name: | family with sequence similarity 19 member A5, C-C motif chemokine like |
Gene Aliases: |
QLLK5208, TAFA-5, TAFA5, UNQ5208 |
Location: |
Chr.22:48489460-48751935 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM19A5 | NM_001082967.2 | NP_001076436.1 | ||
| AF131851.1 | AAD20061.1 | |||
| BC039396.1 | AAH39396.2 | |||
| DB481230.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv778n67 | Chr.22:48453824 - 48657701 on Build GRCh38 | Gain |
|
| nsv829310 | Chr.22:48488182 - 48492339 on Build GRCh38 | Gain |
|
| nsv955186 | Chr.22:48484289 - 48570188 on Build GRCh38 | Deletion |
|
| nsv829311 | Chr.22:48488947 - 48490430 on Build GRCh38 | Gain |
|
| nsv1064218 | Chr.22:48472061 - 48792098 on Build GRCh38 | Gain |
|
| nsv834232 | Chr.22:48358817 - 48553110 on Build GRCh38 | Loss |
|
| nsv829304 | Chr.22:48482687 - 48500457 on Build GRCh38 | Gain |
|
| nsv829305 | Chr.22:48486113 - 48496115 on Build GRCh38 | Gain |
|
| nsv589147 | Chr.22:48310704 - 48549198 on Build GRCh38 | Gain |
|
| dgv779n67 | Chr.22:48486769 - 48492772 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map