Assay Details
Target Gene Details
Entrez Gene ID: | 101927279 |
Gene Name: | EP300 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.22:41185214-41197501 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EP300-AS1 | NR_110514.1 | |||
| AW511192.1 | ||||
| HY041000.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv962797 | Chr.22:41182537 - 41191917 on Build GRCh38 | Duplication |
|
| nsv524585 | Chr.22:40993505 - 41794305 on Build GRCh38 | Loss |
|
| nsv589049 | Chr.22:41168704 - 41276267 on Build GRCh38 | Gain |
|
| esv3575480 | Chr.22:41191014 - 41223706 on Build GRCh38 | Gain |
|

Genomic Map