Assay Details
Target Gene Details
Entrez Gene ID: | 8228 |
Gene Name: | patatin like phospholipase domain containing 4 |
Gene Aliases: |
DXS1283E, GS2, iPLA2eta |
Location: |
Chr.X:7898763-7928587 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PNPLA4 | NM_001142389.1 | NP_001135861.1 | ||
| NM_001172672.1 | NP_001166143.1 | |||
| XM_011545588.1 | 1 | 824 | XP_011543890.1 | |
| BM850121.1 | ||||
| DC421932.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470340 | Chr.X:7857284 - 8463464 on Build GRCh38 | Gain |
|
| esv2752335 | Chr.X:7862723 - 8469952 on Build GRCh38 | Gain |
|
| esv3576759 | Chr.X:7742804 - 8254647 on Build GRCh38 | Gain |
|
| dgv499n21 | Chr.X:7777235 - 8383635 on Build GRCh38 | Gain |
|
| esv2752336 | Chr.X:7904862 - 8063263 on Build GRCh38 | Loss |
|
| esv3576758 | Chr.X:7596710 - 8167603 on Build GRCh38 | Gain |
|
| esv2422194 | Chr.X:7481793 - 8034821 on Build GRCh38 | Duplication |
|
| dgv1n30 | Chr.X:7824572 - 8147412 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map