Assay Details
Target Gene Details
Entrez Gene ID: | 140462 |
Gene Name: | ankyrin repeat and SOCS box containing 9 |
Gene Aliases: |
- |
Location: |
Chr.X:15243987-15270476 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ASB9 | NM_001031739.2 | NP_001026909.1 | ||
| NM_001168530.1 | NP_001162002.1 | |||
| NM_001168531.1 | NP_001162003.1 | |||
| NM_024087.2 | NP_076992.1 | |||
| XM_005274446.1 | XP_005274503.1 | |||
| XM_011545458.1 | XP_011543760.1 | |||
| XM_011545460.1 | XP_011543762.1 | |||
| XM_017029283.1 | XP_016884772.1 | |||
| XM_017029284.1 | XP_016884773.1 | |||
| XM_017029285.1 | XP_016884774.1 | |||
| XM_017029286.1 | XP_016884775.1 | |||
| AK000643.1 | BAA91302.1 | |||
| AK292270.1 | ||||
| AL080091.2 | CAB45706.2 | |||
| AY337578.1 | AAQ17469.1 | |||
| BC001244.1 | AAH01244.1 | |||
| BC013172.1 | AAH13172.1 | |||
| DA943920.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3573602 | Chr.X:15259550 - 15277847 on Build GRCh38 | Loss |
|
| dgv2226e212 | Chr.X:15248328 - 15272357 on Build GRCh38 | Loss |
|
| dgv2227e212 | Chr.X:15259532 - 15272357 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map