Assay Details
Target Gene Details
Entrez Gene ID: | 50814 |
Gene Name: | NAD(P) dependent steroid dehydrogenase-like |
Gene Aliases: |
H105E3, SDR31E1, XAP104 |
Location: |
Chr.X:152830967-152869363 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NSDHL | NM_001129765.1 | NP_001123237.1 | ||
| NM_015922.2 | NP_057006.1 | |||
| XM_011531178.2 | XP_011529480.1 | |||
| XM_017029564.1 | XP_016885053.1 | |||
| AK026549.1 | ||||
| BC000245.1 | AAH00245.1 | |||
| BC007816.2 | AAH07816.1 | |||
| U47105.2 | AAC50558.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv7158 | Chr.X:152841419 - 152868689 on Build GRCh38 | Insertion |
|
| nsv470373 | Chr.X:152796870 - 152935948 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map