Assay Details
Target Gene Details
Entrez Gene ID: | 1756 |
Gene Name: | dystrophin |
Gene Aliases: |
BMD, CMD3B, DXS142, DXS164, DXS206, DXS230, DXS239, DXS268, DXS269, DXS270, DXS272, MRX85 |
Location: |
Chr.X:31119219-33339609 on Build GRCh38 |
Assay Gene Location: | Within Intron 86 |
More Information
Additional Information:
For this assay, SNP(s) [rs140524252] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map