Assay Details
Target Gene Details
Entrez Gene ID: | 340554 |
Gene Name: | zinc finger CCCH-type containing 12B |
Gene Aliases: |
CXorf32, MCPIP2 |
Location: |
Chr.X:65034815-65507887 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZC3H12B | XM_017029478.1 | XP_016884967.1 | ||
| XM_017029479.1 | XP_016884968.1 | |||
| XM_017029480.1 | XP_016884969.1 | |||
| XM_017029481.1 | XP_016884970.1 | |||
| XM_017029482.1 | XP_016884971.1 | |||
| XM_017029483.1 | XP_016884972.1 | |||
| XM_017029484.1 | XP_016884973.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs139230860] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map