Assay Details
Target Gene Details
Entrez Gene ID: | 1756 |
Gene Name: | dystrophin |
Gene Aliases: |
BMD, CMD3B, DXS142, DXS164, DXS206, DXS230, DXS239, DXS268, DXS269, DXS270, DXS272, MRX85 |
Location: |
Chr.X:31119219-33339609 on Build GRCh38 |
Assay Gene Location: | Within Intron 85 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2661363 | Chr.X:31137680 - 31139688 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map