Assay Details
Target Gene Details
Entrez Gene ID: | 5277 |
Gene Name: | phosphatidylinositol glycan anchor biosynthesis class A |
Gene Aliases: |
GPI3, MCAHS2, PIG-A, PNH1 |
Location: |
Chr.X:15319451-15335554 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIGA | NM_002641.3 | NP_002632.1 | ||
| NM_020473.3 | NP_065206.3 | |||
| NR_033835.1 | ||||
| NR_033836.1 | ||||
| XM_011545539.2 | 1 | 320 | XP_011543841.1 | |
| XM_017029581.1 | XP_016885070.1 | |||
| AK098047.1 | ||||
| AK290497.1 | ||||
| AK303538.1 | ||||
| AK304144.1 | ||||
| AK315063.1 | ||||
| BC038236.1 | AAH38236.1 | |||
| CX870731.1 | ||||
| D11466.1 | BAA02019.1 | |||
| DA951065.1 | ||||
| DC347872.1 | ||||
| S74936.1 | AAD14160.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3573603 | Chr.X:15328823 - 15331931 on Build GRCh38 | Loss |
|
| nsv522729 | Chr.X:15283411 - 15334052 on Build GRCh38 | Gain |
|
| nsv517938 | Chr.X:15296656 - 15331048 on Build GRCh38 | Gain |
|
| nsv528343 | Chr.X:15271931 - 15385229 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map