Assay Details
Target Gene Details
Entrez Gene ID: | 139081 |
Gene Name: | MAGE family member C3 |
Gene Aliases: |
CT7.2, HCA2, MAGE-C3, MAGEC4 |
Location: |
Chr.X:141838316-141897832 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAGEC3 | NM_138702.1 | NP_619647.1 | ||
| XM_017029265.1 | XP_016884754.1 | |||
| AF490508.1 | AAM08355.1 | |||
| AK297650.1 | ||||
| BC103893.1 | AAI03894.1 | |||
| BC103894.1 | AAI03895.1 |
More Information
Set Membership: |
|

Genomic Map