Assay Details
Target Gene Details
Entrez Gene ID: | 2986 |
Gene Name: | guanylate cyclase 2F, retinal |
Gene Aliases: |
CYGF, GC-F, GUC2DL, GUC2F, RETGC-2, ROS-GC2 |
Location: |
Chr.X:109372061-109482056 on Build GRCh38 |
Assay Gene Location: | Within Intron 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GUCY2F | NM_001522.2 | NP_001513.2 | ||
| L37378.1 | AAA74451.1 |
More Information
Set Membership: |
|

Genomic Map