Assay Details
Target Gene Details
Entrez Gene ID: | 5638 |
Gene Name: | proline rich and Gla domain 1 |
Gene Aliases: |
PRGP1 |
Location: |
Chr.X:37349275-37457295 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRRG1 | NM_000950.2 | NP_000941.1 | ||
| NM_001142395.1 | NP_001135867.1 | |||
| NM_001173486.1 | NP_001166957.1 | |||
| NM_001173489.1 | NP_001166960.1 | |||
| NM_001173490.1 | NP_001166961.1 | |||
| AF009242.1 | AAB67070.1 | |||
| AK312904.1 | ||||
| BC030786.2 | AAH30786.2 | |||
| BC060833.1 | AAH60833.1 | |||
| DA245329.1 | ||||
| DA411839.1 | ||||
| DA417581.1 | ||||
| DB286340.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv6863 | Chr.X:37318664 - 37351668 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map