Assay Details
Target Gene Details
Entrez Gene ID: | 2187 |
Gene Name: | Fanconi anemia complementation group B |
Gene Aliases: |
FA2, FAAP90, FAAP95, FAB, FACB |
Location: |
Chr.X:14690863-14873255 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FANCB |
Target Gene Details
Entrez Gene ID: | 2742 |
Gene Name: | glycine receptor alpha 2 |
Gene Aliases: |
GLR |
Location: |
Chr.X:14448779-14731812 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GLRA2 | NM_001118885.1 | NP_001112357.1 | ||
| NM_001118886.1 | NP_001112358.1 | |||
| NM_001171942.1 | NP_001165413.1 | |||
| NM_002063.3 | NP_002054.1 | |||
| XM_006724487.3 | XP_006724550.1 | |||
| XM_011545495.2 | XP_011543797.1 | |||
| XM_011545496.2 | XP_011543798.1 | |||
| XM_017029427.1 | XP_016884916.1 | |||
| XM_017029428.1 | XP_016884917.1 | |||
| XM_017029429.1 | XP_016884918.1 | |||
| AK289561.1 | ||||
| AK297268.1 | ||||
| AK297304.1 | ||||
| AK312591.1 | ||||
| AY437083.1 | AAR87842.1 | |||
| AY437084.1 | AAR87843.1 | |||
| BC032864.2 | AAH32864.1 | |||
| CR450343.1 | CAG29339.1 | |||
| X52008.1 | CAA36257.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv502n21 | Chr.X:14591881 - 14702502 on Build GRCh38 | Gain |
|
| nsv526475 | Chr.X:14363314 - 14702502 on Build GRCh38 | Gain |
|
| nsv518002 | Chr.X:14543393 - 14702502 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map