Assay Details
Target Gene Details
Entrez Gene ID: | 139081 |
Gene Name: | MAGE family member C3 |
Gene Aliases: |
CT7.2, HCA2, MAGE-C3, MAGEC4 |
Location: |
Chr.X:141838316-141897832 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAGEC3 | NM_138702.1 | NP_619647.1 | ||
| AF490508.1 | AAM08355.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522089 | Chr.X:141840747 - 141876963 on Build GRCh38 | Gain |
|
| esv3574879 | Chr.X:141858172 - 141865972 on Build GRCh38 | Loss |
|
| esv3574880 | Chr.X:141863497 - 141866211 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map