Assay Details
Target Gene Details
Entrez Gene ID: | 8237 |
Gene Name: | ubiquitin specific peptidase 11 |
Gene Aliases: |
UHX1 |
Location: |
Chr.X:47232915-47248328 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| USP11 | NM_004651.3 | NP_004642.2 | ||
| XM_005272674.3 | XP_005272731.1 | |||
| XM_011543988.1 | XP_011542290.1 | |||
| AB073597.1 | BAC20463.1 | |||
| AK055588.1 | ||||
| AK098394.1 | ||||
| AK294635.1 | ||||
| BC000350.2 | AAH00350.4 | |||
| BC063668.1 | AAH63668.1 | |||
| BC140849.1 | ||||
| U44839.1 | AAC50450.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2740138 | Chr.X:47233705 - 47233860 on Build GRCh38 | Deletion |
|
| esv2740137 | Chr.X:47233454 - 47234183 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map