Assay Details
Target Gene Details
Entrez Gene ID: | 9758 |
Gene Name: | FERM and PDZ domain containing 4 |
Gene Aliases: |
PDZD10, PDZK10 |
Location: |
Chr.X:12137329-12724523 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FRMPD4 | XM_017029983.1 | 1 | 353 | XP_016885472.1 |
| XM_017029986.1 | 1 | 352 | XP_016885475.1 |
More Information
Set Membership: |
|

Genomic Map