Assay Details
Target Gene Details
Entrez Gene ID: | 105373244 |
Gene Name: | uncharacterized LOC105373244 |
Gene Aliases: |
- |
Location: |
Chr.X:70427358-70436800 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105373244 | XM_011531081.2 | 1 | 284 | XP_011529383.1 |
| XM_011531082.2 | 1 | 284 | XP_011529384.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv33082 | Chr.X:70420685 - 71888183 on Build GRCh38 | Loss |
|

Genomic Map