Assay Details
Target Gene Details
Entrez Gene ID: | 158584 |
Gene Name: | fatty acid amide hydrolase 2 |
Gene Aliases: |
AMDD |
Location: |
Chr.X:57121861-57489196 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAAH2 | XM_011530767.2 | XP_011529069.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3558898 | Chr.X:57151781 - 57213067 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map