Assay Details
Target Gene Details
Entrez Gene ID: | 7404 |
Gene Name: | ubiquitously transcribed tetratricopeptide repeat containing, Y-linked |
Gene Aliases: |
KDM6AL, UTY1 |
Location: |
Chr.Y:13230770-13480670 on Build GRCh38 |
Assay Gene Location: | Within Exon 42 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UTY | XM_011531441.2 | 31 | 6346 | XP_011529743.1 |
| XM_011531442.2 | 30 | 6298 | XP_011529744.1 | |
| XM_011531443.2 | 31 | 6280 | XP_011529745.1 | |
| XM_011531445.2 | 30 | 6211 | XP_011529747.1 | |
| XM_011531446.2 | 30 | 6190 | XP_011529748.1 | |
| XM_011531447.2 | 29 | 6163 | XP_011529749.1 | |
| XM_011531448.2 | 30 | 6145 | XP_011529750.1 | |
| XM_011531449.2 | 29 | 6142 | XP_011529751.1 | |
| XM_011531450.2 | 29 | 6097 | XP_011529752.1 | |
| XM_011531451.2 | 29 | 6055 | XP_011529753.1 | |
| XM_011531452.2 | 28 | 6007 | XP_011529754.1 |
More Information
Set Membership: |
|

Genomic Map