Assay Details
Target Gene Details
Entrez Gene ID: | 83259 |
Gene Name: | protocadherin 11 Y-linked |
Gene Aliases: |
PCDH-PC, PCDH22, PCDHX, PCDHY |
Location: |
Chr.Y:5000044-5742228 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCDH11Y | NM_032973.2 | NP_116755.1 | ||
| XM_017030079.1 | XP_016885568.1 | |||
| XM_017030080.1 | XP_016885569.1 | |||
| XM_017030081.1 | XP_016885570.1 | |||
| AF332217.1 | AAK13469.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3577576 | Chr.Y:5026633 - 5785419 on Build GRCh38 | Gain |
|
| esv3577577 | Chr.Y:5439610 - 5723734 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map