Assay Details
Target Gene Details
Entrez Gene ID: | 27245 |
Gene Name: | AT-hook DNA binding motif containing 1 |
Gene Aliases: |
MRD25 |
Location: |
Chr.1:27534245-27604178 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AHDC1 | NM_001029882.3 | NP_001025053.1 | ||
| XM_005245848.3 | XP_005245905.1 | |||
| XM_005245849.3 | XP_005245906.1 | |||
| XM_005245850.3 | XP_005245907.1 | |||
| XM_005245851.3 | XP_005245908.1 | |||
| XM_005245852.3 | XP_005245909.1 | |||
| XM_011541255.1 | XP_011539557.1 | |||
| XM_011541256.2 | XP_011539558.1 | |||
| XM_011541257.2 | XP_011539559.1 | |||
| AK125431.1 | BAC86163.1 | |||
| BC002677.2 | AAH02677.2 | |||
| BC014394.1 | ||||
| U79259.1 | AAB50205.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv520790 | Chr.1:27532525 - 27549971 on Build GRCh38 | Loss |
|
| dgv14n27 | Chr.1:27445496 - 27618848 on Build GRCh38 | Loss |
|
| dgv219n54 | Chr.1:27459823 - 27618848 on Build GRCh38 | Loss |
|
| nsv460917 | Chr.1:27522789 - 27610945 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map