Assay Details
Target Gene Details
Entrez Gene ID: | 10911 |
Gene Name: | urotensin 2 |
Gene Aliases: |
PRO1068, U-II, UCN2, UII |
Location: |
Chr.1:7846345-7913615 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UTS2 | XM_017000116.1 | 7 | 1056 | XP_016855605.1 |
| XM_017000117.1 | 6 | 701 | XP_016855606.1 | |
| XM_017000118.1 | 5 | 508 | XP_016855607.1 | |
| XM_017000119.1 | 6 | 977 | XP_016855608.1 |
More Information
Set Membership: |
|

Genomic Map