Assay Details
Target Gene Details
Entrez Gene ID: | 84919 |
Gene Name: | protein phosphatase 1 regulatory subunit 15B |
Gene Aliases: |
CREP, MSSGM2 |
Location: |
Chr.1:204398684-204411880 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPP1R15B | XM_005245551.4 | 3 | 3240 | XP_005245608.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522489 | Chr.1:204396621 - 204401286 on Build GRCh38 | Loss |
|
| nsv952641 | Chr.1:204396873 - 204410972 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map