Assay Details
Target Gene Details
Entrez Gene ID: | 23038 |
Gene Name: | WD and tetratricopeptide repeats 1 |
Gene Aliases: |
ADP, DCAF9 |
Location: |
Chr.1:27234516-27308633 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WDTC1 | NM_001276252.1 | NP_001263181.1 | ||
| NM_015023.4 | NP_055838.2 | |||
| XM_011541057.1 | XP_011539359.1 | |||
| XM_011541058.2 | XP_011539360.1 | |||
| XM_017000730.1 | XP_016856219.1 | |||
| AK001734.1 | BAA91868.1 | |||
| AK315564.1 | ||||
| BC032523.1 | AAH32523.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1010793 | Chr.1:27214723 - 27282027 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map