Assay Details
Target Gene Details
Entrez Gene ID: | 79573 |
Gene Name: | tetratricopeptide repeat domain 13 |
Gene Aliases: |
- |
Location: |
Chr.1:230906241-230978872 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTC13 | NM_001122835.2 | NP_001116307.2 | ||
| NM_024525.4 | NP_078801.3 | |||
| XM_005273260.1 | XP_005273317.1 | |||
| XM_005273261.2 | XP_005273318.1 | |||
| XM_005273262.1 | XP_005273319.1 | |||
| XM_005273264.2 | XP_005273321.1 | |||
| XM_006711814.2 | XP_006711877.1 | |||
| XM_006711815.2 | XP_006711878.1 | |||
| XM_011544275.2 | XP_011542577.1 | |||
| XM_011544276.2 | XP_011542578.1 | |||
| XM_017002321.1 | XP_016857810.1 | |||
| XM_017002322.1 | XP_016857811.1 | |||
| XM_017002323.1 | XP_016857812.1 | |||
| XM_017002324.1 | XP_016857813.1 | |||
| XM_017002325.1 | XP_016857814.1 | |||
| XM_017002326.1 | XP_016857815.1 | |||
| AK075386.1 | BAC11586.1 | |||
| AK075561.1 | BAC11700.1 | |||
| AK309725.1 | ||||
| AL832286.1 | CAH10599.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3589093 | Chr.1:230482037 - 231277190 on Build GRCh38 | Gain |
|
| nsv1000488 | Chr.1:230799658 - 231269735 on Build GRCh38 | Gain |
|

Genomic Map