Assay Details
Target Gene Details
Entrez Gene ID: | 80143 |
Gene Name: | suppressor of IKBKE 1 |
Gene Aliases: |
SIKE |
Location: |
Chr.1:114769479-114780687 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SIKE1 | NM_001102396.1 | 5 | 3180 | NP_001095866.1 |
| NM_025073.2 | 5 | 3168 | NP_079349.2 | |
| NR_049741.1 | 5 | 3230 | ||
| NR_049742.1 | 4 | 3062 | ||
| AK126316.1 | 5 | 3178 | ||
| CR749219.1 | 5 | 3171 | CAH18076.1 | |
| CR936771.1 | 5 | 3171 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv28734 | Chr.1:114376708 - 115005298 on Build GRCh38 | Loss |
|
| nsv824265 | Chr.1:114769762 - 114774342 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map