Assay Details
Target Gene Details
Entrez Gene ID: | 23380 |
Gene Name: | SLIT-ROBO Rho GTPase activating protein 2 |
Gene Aliases: |
ARHGAP34, FNBP2, SRGAP2A, SRGAP3 |
Location: |
Chr.1:206203464-206464443 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SRGAP2 | NM_001170637.3 | NP_001164108.1 | ||
| NM_001300952.1 | NP_001287881.1 | |||
| NM_015326.4 | NP_056141.2 | |||
| XM_005277510.2 | XP_005277567.1 | |||
| XM_005277511.2 | XP_005277568.1 | |||
| XM_005277514.1 | XP_005277571.1 | |||
| XM_005277515.1 | XP_005277572.1 | |||
| XM_011509354.1 | XP_011507656.1 | |||
| XM_011509355.1 | XP_011507657.1 | |||
| XM_011509356.2 | XP_011507658.1 | |||
| XM_011509359.1 | XP_011507661.1 | |||
| XM_017000838.1 | XP_016856327.1 | |||
| XM_017000839.1 | XP_016856328.1 | |||
| XM_017000840.1 | XP_016856329.1 | |||
| XM_017000841.1 | XP_016856330.1 | |||
| AB007925.1 | ||||
| AK293335.1 | ||||
| AK295845.1 | ||||
| BC132872.1 | ||||
| BC132874.1 | ||||
| BC144343.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv946612 | Chr.1:206309524 - 206410530 on Build GRCh38 | Duplication |
|
| esv28559 | Chr.1:206382600 - 206430060 on Build GRCh38 | Gain |
|
| nsv821120 | Chr.1:206380831 - 206410817 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map