Assay Details
Target Gene Details
Entrez Gene ID: | 100505633 |
Gene Name: | long intergenic non-protein coding RNA 1133 |
Gene Aliases: |
- |
Location: |
Chr.1:159961224-159979086 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LINC01133 | NR_038849.1 | |||
| AF088076.1 | ||||
| CB119247.1 |
Target Gene Details
Entrez Gene ID: | 89886 |
Gene Name: | SLAM family member 9 |
Gene Aliases: |
CD2F-10, CD2F10, CD84-H1, CD84H1, SF2001 |
Location: |
Chr.1:159951492-159984730 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLAMF9 | XM_017002756.1 | XP_016858245.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv548055 | Chr.1:159948628 - 159970210 on Build GRCh38 | Loss |
|
| nsv520773 | Chr.1:159953169 - 159965779 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map